STF-118804 ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-(5-Methyl-4-(tosylmethyl)oxazol-2-yl)-N-(pyridin-3-ylmethyl)benzamide

A692427-10MG A692427-1MG A692427-5MG
77117-888EA 22.28 USD
77117-888 77117-890 77117-892
STF-118804 ≥98%
STF-118804

Precaución: for Research Use Only

Formula: C₂₅H₂₃N₃O₄S
MW: 461.54 g/mol
MDL Number: MFCD14918065
CAS Number: 894187-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR