1,6-Dimethoxynaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dimethoxynaphthalene

A689104-100G A689104-1G A689104-25G A689104-5G
77117-878EA 349.04 USD
77117-878 77117-880 77117-882 77117-884
1,6-Dimethoxynaphthalene 98%
1,6-Dimethoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
MDL Number: MFCD00086330
CAS Number: 3900-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR