3-Chloro-4-fluorobenzoyl chloride 98%

Supplier: Ambeed
Danger

A685349-1G A685349-25G A685349-5G
77117-866EA 10.31 USD
77117-866 77117-868 77117-870
3-Chloro-4-fluorobenzoyl chloride 98%
3-Chloro-4-fluorobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO
MDL Number: MFCD00236215
CAS Number: 65055-17-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR