1,3-Dithiane 98%

Supplier: Ambeed
Danger

A684474-100G A684474-10G A684474-1G A684474-25G A684474-5G
77117-856EA 155.54 USD
77117-856 77117-858 77117-860 77117-862 77117-864
1,3-Dithiane 98%
1,3-Dithiane

Precaución: for Research Use Only

Formula: C₄H₈S₂
MDL Number: MFCD00006654
CAS Number: 505-23-7
UN: 3335
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR