MK-2461 98+%

Supplier: Ambeed
Warning

Synonyms: N-((2R)-1,4-Dioxan-2-ylmethyl)-N-methyl-N'-[3-(1-methyl-1H-pyrazol-4-yl)-5-oxo-5H-benzo[4,5]cyclohepta[1,2-b]pyridin-7-yl]sulphamide

A683250-1MG A683250-2MG A683250-5MG
77117-838EA 82.76 USD
77117-838 77117-840 77117-842
MK-2461 98+%
MK-2461

Precaución: for Research Use Only

Formula: C₂₄H₂₅N₅O₅S
MW: 495.6 g/mol
MDL Number: MFCD11977739
CAS Number: 917879-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR