6-Methoxy-3,4-dihydro-2H-1,4-benzoxazine 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-3,4-dihydro-2H-benzo[b][1,4]oxazine

A683238-250MG A683238-100MG A683238-1G
77117-836EA 119.89 USD
77117-836 77686-655 77686-656
6-Methoxy-3,4-dihydro-2H-1,4-benzoxazine 95%
6-Methoxy-3,4-dihydro-2H-1,4-benzoxazine

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.189 g/mol
MDL Number: MFCD08236770
CAS Number: 58960-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR