N-Ethyl-3-bromobenzylamine 97%

Supplier: Ambeed
Danger

A683057-1G A683057-25G A683057-5G
77117-830EA 33.95 USD
77117-830 77117-832 77117-834
N-Ethyl-3-bromobenzylamine 97%
N-Ethyl-3-bromobenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
MW: 214.10 g/mol
MDL Number: MFCD07405019
CAS Number: 90389-91-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR