tert-Butyl [4-(methylamino)butyl]carbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-(methylamino)butyl)carbamate

A682832-100MG A682832-1G A682832-250MG A682832-5G
77117-816EA 20.6 USD
77117-816 77117-818 77117-820 77686-653
tert-Butyl [4-(methylamino)butyl]carbamate 97%
tert-Butyl [4-(methylamino)butyl]carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₂N₂O₂
MDL Number: MFCD06808587
CAS Number: 874831-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR