1,2-Bis(pyridin-4-ylmethylene)hydrazine 95%

Supplier: Ambeed
Warning

A680642-100MG A680642-250MG A680642-5G
77117-808EA 31.83 USD
77117-808 77117-810 77117-812
1,2-Bis(pyridin-4-ylmethylene)hydrazine 95%
1,2-Bis(pyridin-4-ylmethylene)hydrazine

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₄
MW: 210.23 g/mol
MDL Number: MFCD00136124
CAS Number: 6957-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR