1-Ethyl-4-piperidone 98%

Supplier: Ambeed
Warning

A677434-100G A677434-10G A677434-1G A677434-25G A677434-5G
77117-798EA 110.21 USD
77117-798 77117-800 77117-802 77117-804 77117-806
1-Ethyl-4-piperidone 98%
1-Ethyl-4-piperidone

Precaución: for Research Use Only

Formula: C₇H₁₃NO
MDL Number: MFCD00013356
CAS Number: 3612-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR