Bromodifluoroacetic acid 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Bromodifluoroaceticacid
A677254-25G
A677254-5G
A677254-100G
77117-796EA
31.83
USD
77117-796
77569-286
77686-646
Bromodifluoroacetic acid 98%
Bromodifluoroacetic acid
Precaución: For research use only.
Formula:
C₂HBrF₂O₂ MW: 174.93 g/mol |
MDL Number:
MFCD02183519 CAS Number: 354-08-5 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...