Sodium 2,3,4,5-tetrafluorobenzoate 95%

Supplier: Ambeed
Warning

A677115-100G A677115-10G A677115-25G
77117-790EA 160.2 USD
77117-790 77117-792 77117-794
Sodium 2,3,4,5-tetrafluorobenzoate 95%
Sodium 2,3,4,5-tetrafluorobenzoate

Precaución: for Research Use Only

Formula: C₇HF₄NaO₂
MW: 216.06 g/mol
MDL Number: MFCD03094400
CAS Number: 67852-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR