4-Pyridinethiol 97%

Supplier: Ambeed
Warning

Synonyms: Pyridine-4-thiol, 4-Mercaptopyridine

A676901-1G A676901-25G A676901-5G
77117-784EA 6.37 USD
77117-784 77117-786 77117-788
4-Pyridinethiol 97%
4-Pyridinethiol

Precaución: for Research Use Only

Formula: C₅H₅NS
MW: 111.17 g/mol
MDL Number: MFCD00006422
CAS Number: 4556-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR