2-Chlorothiophenol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
o-Chlorothiophenol, 2-chlorothiophenol, 1-Chloro-2-mercaptobenzene, 2-Chlorobenzenethiol, 2-Chlorobenzene-1-thiol
A673368-25G
A673368-5G
A673368-10G
77117-776EA
29.71
USD
77117-776
77117-778
77569-280
2-Chlorothiophenol 98%
2-Chlorothiophenol
Precaución: For research use only.
Formula:
C₆H₅ClS MW: 144.62 g/mol |
MDL Number:
MFCD00004830 CAS Number: 6320-03-2 UN: 3265 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...