3-Hydroxybenzyl alcohol 97%

Supplier: Ambeed
Warning

A673062-100G A673062-10G A673062-1G A673062-25G A673062-5G
77117-766EA 100.94 USD
77117-766 77117-768 77117-770 77117-772 77117-774
3-Hydroxybenzyl alcohol 97%
3-Hydroxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₈O₂
MDL Number: MFCD00004643
CAS Number: 620-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR