2-((5-Methoxy-1H-indol-3-yl)methylene)-N-pentylhydrazine-1-carboximidamide maleate 98%

Supplier: Ambeed
Warning

A670060-10MG A670060-50MG
77117-758EA 5.31 USD
77117-758 77117-760
2-((5-Methoxy-1H-indol-3-yl)methylene)-N-pentylhydrazine-1-carboximidamide maleate 98%
2-((5-Methoxy-1H-indol-3-yl)methylene)-N-pentylhydrazine-1-carboximidamide maleate

Precaución: for Research Use Only

Formula: C₂₀H₂₇N₅O₅
MW: 417.46 g/mol
MDL Number: MFCD04116201
CAS Number: 189188-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR