6-Chloro-4-hydroxy-8-methylquinoline-3-carboxylic acid ethyl ester 98+%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-chloro-4-hydroxy-8-methylquinoline-3-carboxylate

A668596-100MG A668596-1G A668596-250MG
77117-752EA 31.83 USD
77117-752 77117-754 77117-756
6-Chloro-4-hydroxy-8-methylquinoline-3-carboxylic acid ethyl ester 98+%
6-Chloro-4-hydroxy-8-methylquinoline-3-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNO₃
MW: 265.692 g/mol
MDL Number: MFCD00173359
CAS Number: 228728-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR