1-Chloro-3,5-dibromo-4-fluorobenzene 98%

Supplier: Ambeed
Warning

A668166-1G A668166-250MG A668166-5G
77117-746EA 57.29 USD
77117-746 77117-748 77117-750
1-Chloro-3,5-dibromo-4-fluorobenzene 98%
1-Chloro-3,5-dibromo-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂ClF
MW: 288.339 g/mol
MDL Number: MFCD28166159
CAS Number: 1805525-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR