(S)-5-Phenylmorpholin-3-one 95%

Supplier: Ambeed
Warning

A667548-100MG A667548-1G A667548-250MG
77117-740EA 15.45 USD
77117-740 77117-742 77117-744
(S)-5-Phenylmorpholin-3-one 95%
(S)-5-Phenylmorpholin-3-one

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD19442818
CAS Number: 1052209-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR