DL-2-Aminopimelic acid 98%

Supplier: Ambeed
Warning

Synonyms: DL-2-Aminoheptanedioic acid, 2-Aminoheptanedioic acid

A666316-100MG A666316-1G A666316-250MG A666316-5G
77117-730EA 16.98 USD
77117-730 77117-732 77117-734 77117-736
DL-2-Aminopimelic acid 98%
DL-2-Aminopimelic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₄
MW: 175.18 g/mol
MDL Number: MFCD00053314
CAS Number: 627-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR