4-Hydroxy-2-(trifluoromethyl)benzamide 98%

Supplier: Ambeed
Warning

A665609-1G A665609-25G A665609-5G
77117-722EA 18.04 USD
77117-722 77117-724 77117-726
4-Hydroxy-2-(trifluoromethyl)benzamide 98%
4-Hydroxy-2-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MW: 205.13 g/mol
MDL Number: MFCD13176522
CAS Number: 1208077-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR