1-Bromo-2,4-dimethoxy-5-fluorobenzene 95%

Supplier: Ambeed
Warning

A662378-100MG A662378-250MG
77117-710EA 27.81 USD
77117-710 77117-712
1-Bromo-2,4-dimethoxy-5-fluorobenzene 95%
1-Bromo-2,4-dimethoxy-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
MDL Number: MFCD12547863
CAS Number: 1208077-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR