Guanidinium carbonate 99%

Supplier: Ambeed
Danger

Synonyms: Guanidine carbonate salt, Diguanidinium carbonate, Guanidine carbonate

A661494-25G A661494-100G
77117-708EA 6.18 USD
77117-708 77569-256
Guanidinium carbonate 99%
Guanidinium carbonate

Precaución: For research use only.

Formula: C₃H₁₂N₆O₃
MW: 180.17 g/mol
MDL Number: MFCD00013029
CAS Number: 593-85-1
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR