2,5-Dibromothiazole 97%

Supplier: Ambeed
Warning

A661115-1G A661115-25G A661115-5G A661115-100G
77117-702EA 9.27 USD
77117-702 77117-704 77117-706 77617-764
2,5-Dibromothiazole 97%
2,5-Dibromothiazole

Precaución: for Research Use Only

Formula: C₃HBr₂NS
MDL Number: MFCD00016891
CAS Number: 4175-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR