2-Chloro-4-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A655004-100G A655004-10G A655004-25G A655004-500G A655004-5G
77117-684EA 38.2 USD
77117-684 77117-686 77117-688 77117-690 77117-692
2-Chloro-4-fluorobenzoic acid 98%
2-Chloro-4-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
MDL Number: MFCD00010615
CAS Number: 2252-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR