1-Chloro-4-ethynylbenzene 97%

Supplier: Ambeed
Danger

Synonyms: (4-Chlorophenyl)acetylene, 1-Ethynyl-4-chlorobenzene, (4-Chlorophenyl)ethyne

A651662-100MG A651662-10G A651662-1G A651662-250MG A651662-25G A651662-5G
77117-656EA 5.15 USD
77117-656 77117-658 77117-660 77117-662 77117-664 77117-666
1-Chloro-4-ethynylbenzene 97%
1-Chloro-4-ethynylbenzene

Precaución: for Research Use Only

Formula: C₈H₅Cl
MDL Number: MFCD00191917
CAS Number: 873-73-4
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR