2-Ethoxy-3-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Ethoxypyridin-3-yl)boronic acid

A651143-1G A651143-5G A651143-25G
77117-644EA 7.21 USD
77117-644 77117-646 77614-018
2-Ethoxy-3-pyridineboronic acid 98%
2-Ethoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₃
MDL Number: MFCD06201036
CAS Number: 854373-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR