4-Fluorobenzophenone 98%

Supplier: Ambeed
Warning

A647785-100G A647785-25G A647785-500G A647785-5G
77117-626EA 22.28 USD
77117-626 77117-628 77117-630 77117-632
4-Fluorobenzophenone 98%
4-Fluorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₉FO
MW: 200.21 g/mol
MDL Number: MFCD00000352
CAS Number: 345-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR