2-Fluoro-5-iodoaniline 98%

Supplier: Ambeed
Warning

A641939-1G A641939-250MG A641939-25G A641939-5G
77117-586EA 18.55 USD
77117-586 77117-588 77117-590 77613-990
2-Fluoro-5-iodoaniline 98%
2-Fluoro-5-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅FIN
MDL Number: MFCD06796596
CAS Number: 886362-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR