(4-Bromophenyl)hydrazine xhydrochloride 98%

Supplier: Ambeed
Danger

A641667-1G A641667-25G A641667-5G
77117-580EA 5.31 USD
77117-580 77117-582 77117-584
(4-Bromophenyl)hydrazine xhydrochloride 98%
(4-Bromophenyl)hydrazine xhydrochloride

Precaución: for Research Use Only

Formula: C₆H₈BrClN₂
MW: 223.50 g/mol
CAS Number: 41931-18-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR