N1,N2-Bis(furan-2-ylmethyl)oxalamide 95%

Supplier: Ambeed
Danger

A641381-100G A641381-1G A641381-250MG A641381-25G A641381-5G
77117-570EA 502.87 USD
77117-570 77117-572 77117-574 77117-576 77117-578
N1,N2-Bis(furan-2-ylmethyl)oxalamide 95%
N1,N2-Bis(furan-2-ylmethyl)oxalamide

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₄
MW: 248.23 g/mol
MDL Number: MFCD00090454
CAS Number: 69010-90-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR