2-Bromo-4-fluoro-N-methylbenzamide 95+%

Supplier: Ambeed
Warning

A640981-1G A640981-250MG A640981-5G
77117-564EA 22.28 USD
77117-564 77117-566 77117-568
2-Bromo-4-fluoro-N-methylbenzamide 95+%
2-Bromo-4-fluoro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrFNO
MW: 232.05 g/mol
MDL Number: MFCD08895950
CAS Number: 923138-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR