2-(Morpholinomethyl)-5-((5-((7-(trifluoromethyl)quinolin-4-yl)thio)pentyl)oxy)-4H-pyran-4-one dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: EHT 1864

A638191-10MG A638191-1MG A638191-5MG
77117-552EA 124.13 USD
77117-552 77117-554 77117-556
2-(Morpholinomethyl)-5-((5-((7-(trifluoromethyl)quinolin-4-yl)thio)pentyl)oxy)-4H-pyran-4-one dihydrochloride 98%
2-(Morpholinomethyl)-5-((5-((7-(trifluoromethyl)quinolin-4-yl)thio)pentyl)oxy)-4H-pyran-4-one dihydrochloride

Precaución: for Research Use Only

Formula: C₂₅H₂₉Cl₂F₃N₂O₄S
MW: 581.47 g/mol
MDL Number: MFCD11114384
CAS Number: 754240-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR