1,3-Difluoro-2-isopropoxybenzene 95+%

Supplier: Ambeed
Warning

Synonyms: 1,3-Difluoro-2-(1-methylethoxy)benzene

A638085-1G A638085-250MG
77117-548EA 83.82 USD
77117-548 77117-550
1,3-Difluoro-2-isopropoxybenzene 95+%
1,3-Difluoro-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O
MW: 172.17 g/mol
MDL Number: MFCD22056750
CAS Number: 1219020-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR