1-(2-Chloroethyl)piperidinium chloride 97%

Supplier: Ambeed
Danger

Synonyms: N-(2-Chloroethyl)-piperidine hydrochloride, 1-(2-Chloroethyl)piperidine hydrochloride

A636488-10G A636488-25G
77117-544EA 8.24 USD
77117-544 77117-546
1-(2-Chloroethyl)piperidinium chloride 97%
1-(2-Chloroethyl)piperidinium chloride

Precaución: for Research Use Only

Formula: C₇H₁₅Cl₂N
MW: 184.11 g/mol
MDL Number: MFCD00012837
CAS Number: 2008-75-5
UN: 2928
ADR: 6.1,8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR