WZ-3146 98+%

Supplier: Ambeed
Warning

Synonyms: N-(3-((5-chloro-2-((4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)oxy)phenyl)acrylamide

A635130-10MG A635130-1MG A635130-25MG A635130-5MG A635130-50MG A635130-250MG
77117-534EA 45.62 USD
77117-534 77117-536 77117-538 77117-540 77613-968 77686-582
WZ-3146 98+%
WZ-3146

Precaución: for Research Use Only

Formula: C₂₄H₂₅ClN₆O₂
MW: 464.95 g/mol
MDL Number: MFCD18074503
CAS Number: 1214265-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR