1-(Bromomethyl)-4-fluoro-2-iodobenzene 97%

Supplier: Ambeed
Danger

A631685-100MG A631685-250MG
77117-524EA 28.65 USD
77117-524 77117-526
1-(Bromomethyl)-4-fluoro-2-iodobenzene 97%
1-(Bromomethyl)-4-fluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
MW: 314.92 g/mol
MDL Number: MFCD08543943
CAS Number: 70931-59-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR