2,3-Difluorotoluene 98%

Supplier: Ambeed
Danger

A630049-10G A630049-1G A630049-25G A630049-5G
77117-516EA 24.41 USD
77117-516 77117-518 77117-520 77117-522
2,3-Difluorotoluene 98%
2,3-Difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₆F₂
MW: 128.12 g/mol
MDL Number: MFCD01631506
CAS Number: 3828-49-7
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR