Pyridinium chloride 95%

Supplier: Ambeed
Warning

Synonyms: Pyridine hydrochloride, Pyridinehydrochloride, Pyridine HCl

A629812-1000G A629812-100G A629812-25G A629812-500G
77117-508EA 58.72 USD
77117-508 77117-510 77117-512 77117-514
Pyridinium chloride 95%
Pyridinium chloride

Precaución: for Research Use Only

Formula: C₅H₆ClN
MDL Number: MFCD00012802
CAS Number: 628-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR