Perindopril t-Butylamine Salt 99%

Supplier: Ambeed
Warning

Synonyms: 2-Methylpropan-2-amine (2S,3aS,7aS)-1-((S)-2-(((S)-1-ethoxy-1-oxopentan-2-yl)amino)propanoyl)octahydro-1H-indole-2-carboxylate, (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylic acid tert-butylamine salt, Perindopril tert-butylamine, Perindopril erbumine

A628131-100MG A628131-1G A628131-1MG A628131-250MG A628131-25MG A628131-5G
77117-488EA 14.86 USD
77117-488 77117-490 77117-492 77117-494 77117-496 77117-498
Perindopril t-Butylamine Salt 99%
Perindopril t-Butylamine Salt

Precaución: for Research Use Only

Formula: C₁₉H₃₂N₂O5₅·C₄H₁₁N
MW: 441.6 g/mol
MDL Number: MFCD02313824
CAS Number: 107133-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR