(Bromomethyl)cyclohexane 98%

Supplier: Ambeed
Danger

A627482-1G A627482-25G
77117-484EA 9.26 USD
77117-484 77117-486 77551-888
(Bromomethyl)cyclohexane 98%
(Bromomethyl)cyclohexane

Precaución: for Research Use Only

Formula: C₇H₁₃Br
MDL Number: MFCD00001509
CAS Number: 2550-36-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR