2-Hydroxybenzaldehyde Phenylhydrazone 98%

Supplier: Ambeed
Danger

Synonyms: Salicylaldehyde Phenylhydrazone, 2-((2-Phenylhydrazono)methyl)phenol

A625061-25G A625061-5G
77117-480EA 54.59 USD
77117-480 77117-482
2-Hydroxybenzaldehyde Phenylhydrazone 98%
2-Hydroxybenzaldehyde Phenylhydrazone

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O
MDL Number: MFCD00136159
CAS Number: 614-65-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR