1-Benzoylpiperidine 98%

Supplier: Ambeed
Danger

Synonyms: Phenyl(piperidin-1-yl)methanone

A620991-1G A620991-250MG A620991-25G A620991-5G
77117-454EA 36.08 USD
77117-454 77117-456 77117-458 77117-460
1-Benzoylpiperidine 98%
1-Benzoylpiperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO
MW: 189.26 g/mol
MDL Number: MFCD00023702
CAS Number: 776-75-0
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR