2-Chloro-3-formyl-6-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-methylnicotinaldehyde, 2-Chloro-6-methyl-3-pyridinecarboxaldehyde

A619212-10G A619212-1G A619212-5G
77117-444EA 189.91 USD
77117-444 77117-446 77117-448
2-Chloro-3-formyl-6-picoline 98%
2-Chloro-3-formyl-6-picoline

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MW: 155.5816 g/mol
MDL Number: MFCD07368878
CAS Number: 91591-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR