Dipyridamole 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Bis(diethanolamine)-4,8-dipiperidinopyrimido[5,4-d]pyrimidine, 2,2',2'',2'''-((4,8-Di(piperidin-1-yl)pyrimido[5,4-d]pyrimidine-2,6-diyl)bis(azanetriyl))tetrakis(ethan-1-ol), 2,2',2'',2'''-[(4,8-Dipiperidinylpyrimido[5,4-d]pyrimidine-2,6-diyl)dinitrilo]tetrakisethanol

A619148-100G A619148-10G A619148-1G A619148-25G A619148-5G
77117-438EA 131.56 USD
77117-438 77117-440 77117-442 77613-930 77686-557
Dipyridamole 98%
Dipyridamole

Precaución: for Research Use Only

Formula: C₂₄H₄₀N₈O₄
MW: 504.64 g/mol
MDL Number: MFCD00010555
CAS Number: 58-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR