4-Iodophenylhydrazine 95%

Supplier: Ambeed
Danger

Synonyms: (4-Iodophenyl)hydrazine

A618610-1G A618610-250MG A618610-5G
77117-432EA 46.35 USD
77117-432 77117-434 77117-436
4-Iodophenylhydrazine 95%
4-Iodophenylhydrazine

Precaución: for Research Use Only

Formula: C₆H₇IN₂
MDL Number: MFCD00052688
CAS Number: 13116-27-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR