α'-Chloro-α,α,α-trifluoro-p-xylene 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-(Chloromethyl)benzotrifluoride, 1-(Chloromethyl)-4-(trifluoromethyl)benzene, 1-(Chlormethyl)-4-(trifluormethyl)benzene, 4-(Trifluoromethyl)benzyl chloride
A617984-25G
A617984-5G
77117-422EA
33.95
USD
77117-422
77117-424
α'-Chloro-α,α,α-trifluoro-p-xylene 98%
α'-Chloro-α,α,α-trifluoro-p-xylene
Precaución: for Research Use Only
Formula:
C₈H₆ClF₃ MW: 194.58 g/mol |
MDL Number:
MFCD00040772 CAS Number: 939-99-1 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...