3,5-Dimethyl-1H-pyrazole-1-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethylpyrazole-1-carboxamide, 1-Carbamoyl-3,5-dimethylpyrazole

A617821-1G A617821-25G A617821-5G
77117-408EA 10.61 USD
77117-408 77117-410 77117-412
3,5-Dimethyl-1H-pyrazole-1-carboxamide 98%
3,5-Dimethyl-1H-pyrazole-1-carboxamide

Precaución: for Research Use Only

Formula: C₆H₉N₃O
MW: 139.16 g/mol
MDL Number: MFCD00003126
CAS Number: 934-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR