α-Chloro-3,4-difluorotoulene 98%

Supplier: Ambeed
Danger

A616693-25G A616693-5G
77117-404EA 48.41 USD
77117-404 77117-406
α-Chloro-3,4-difluorotoulene 98%
α-Chloro-3,4-difluorotoulene

Precaución: for Research Use Only

Formula: C₇H₅ClF₂
MDL Number: MFCD00236218
CAS Number: 698-80-6
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR