AMG-208 98+%

Supplier: Ambeed
Warning

A611009-10MG A611009-1MG A611009-25MG A611009-5MG
77117-378EA 40.18 USD
77117-378 77117-380 77117-382 77117-384
AMG-208 98+%
AMG-208

Precaución: for Research Use Only

Formula: C₂₂H₁₇N₅O₂
MDL Number: MFCD17215205
CAS Number: 1002304-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR